AC5372910427 Accela ChemBio Inc.

2-Aminoethanesulfonamide

  • MDL number MFCD08061825
  • Cas Number : 4378-70-5
  • Molecular Weight : 124.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008704&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY008704.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart