AC5372910443 Accela ChemBio Inc.

tert-Butyl Bicyclo[2.2.1]-5-heptene-2-carboxylate

  • MDL number MFCD18083704
  • Cas Number : 154970-45-3
  • Molecular Weight : 194.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008848&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P264-P280-P302+P352+P332+P313+P362+P364-P305+P351+P338+P337+P313-P370+P378
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY008848.pdf
hstatements H227-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart