AC5372910458 Accela ChemBio Inc.

(S)-2-(3-Fluorophenyl)pyrrolidine D-Tartrate

  • MDL number MFCD17171375
  • Cas Number : 1355239-03-0
  • Molecular Weight : 315.2942232

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY008915&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY008915.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart