AC5372910467 Accela ChemBio Inc.

1-Bromo-2-fluoro-4-(methylsulfonyl)benzene

  • MDL number MFCD11053812
  • Cas Number : 1032825-02-7
  • Molecular Weight : 253.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009045&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009045.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart