AC5372910510 Accela ChemBio Inc.

3-(4-Bromophenyl)-1,1,1-trifluoro-2-propanol

  • MDL number MFCD16615965
  • Cas Number : 1148050-30-9
  • Molecular Weight : 269.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009466&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009466.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart