AC5372910525 Accela ChemBio Inc.

Methyl 1-Benzyl-6-oxopiperidine-3-carboxylate

  • MDL number MFCD16883082
  • Cas Number : 156779-11-2
  • Molecular Weight : 247.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009585&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009585.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart