AC5372910535 Accela ChemBio Inc.

Sodium D-Pantothenate

  • MDL number MFCD00002767
  • Cas Number : 867-81-2
  • Molecular Weight : 241.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009652&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY009652.pdf
hstatements H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart