AC5372910542 Accela ChemBio Inc.

2,3,5-Trifluoro-4-methylbenzoic Acid

  • MDL number MFCD00456800
  • Cas Number : 773869-50-4
  • Molecular Weight : 190.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY009676&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY009676.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart