AC5372910579 Accela ChemBio Inc.

Ethyl (R)-Pyrrolidine-3-carboxylate

  • MDL number MFCD18375249
  • Cas Number : 1807380-83-1
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010181&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY010181.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart