AC5372910586 Accela ChemBio Inc.

5-Boc-2-(dimethylamino)-5,6,7,8-tetrahydropyrido[3,2-d]pyrimidine

  • MDL number MFCD17171379
  • Cas Number : 1246471-35-1
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010346&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY010346.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart