AC5372910596 Accela ChemBio Inc.

(4R,6S)-6-Methyl-7,7-dioxo-5,6-dihydro-4H-thieno[2,3-b]thiopyran-4-ol

  • MDL number MFCD11035925
  • Cas Number : 147128-77-6
  • Molecular Weight : 218.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010499&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY010499.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart