AC5372910606 Accela ChemBio Inc.

1-(2-Pyridyl)cyclopropylamine Dihydrochloride

  • MDL number MFCD09028134
  • Cas Number : 1215107-39-3
  • Molecular Weight : 207.10028

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010720&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY010720.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart