AC5372910610 Accela ChemBio Inc.

(S)-2-(3-Pyrrolidinyl)-2-propanol Hydrochloride

  • MDL number MFCD17171384
  • Cas Number : 1273577-45-9
  • Molecular Weight : 165.66104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010744&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY010744.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart