AC5372910618 Accela ChemBio Inc.

3-Fluorodibenz[b,e]oxepin-11(6H)-one

  • MDL number MFCD12828511
  • Cas Number : 114312-48-0
  • Molecular Weight : 228.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY010885&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY010885.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart