AC5372910636 Accela ChemBio Inc.

2-(2-Bromo-6-nitrophenyl)acetic Acid

  • MDL number MFCD10566470
  • Cas Number : 37777-74-5
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011011&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011011.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart