AC5372910640 Accela ChemBio Inc.

3-Chloro-4-methoxyaniline Hydrochloride

  • MDL number MFCD18070993
  • Cas Number : 6329-90-4
  • Molecular Weight : 194.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011022&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011022.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart