AC5372910643 Accela ChemBio Inc.

4-Methyl-2,1,3-benzoxadiazole

  • MDL number MFCD00100600
  • Cas Number : 29091-40-5
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid, or Colorless to yellow liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011026&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P302+P352-P280-P305+P351+P338
MSDS http://pdf.accelachem.com/cn/msds/SY011026.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart