AC5372910688 Accela ChemBio Inc.

4-(4-Bromo-2-fluorophenyl)morpholine

  • MDL number MFCD11846942
  • Cas Number : 513068-89-8
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011575&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011575.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart