AC5372910693 Accela ChemBio Inc.

3-Amino-5-chloro-2-hydroxypyridine

  • MDL number MFCD07375031
  • Cas Number : 98027-36-2
  • Molecular Weight : 144.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011625&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011625.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart