AC5372910699 Accela ChemBio Inc.

Ethyl 3-Aminopyridazine-4-carboxylate

  • MDL number MFCD18071011
  • Cas Number : 716325-02-9
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011639&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011639.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart