AC5372910711 Accela ChemBio Inc.

(R)-3-(4-Fluorophenoxy)pyrrolidine

  • MDL number MFCD11858553
  • Cas Number : 900512-41-6
  • Molecular Weight : 181.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011690&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011690.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart