AC5372910726 Accela ChemBio Inc.

1-(4-Fluorophenyl)-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD02933744
  • Cas Number : 24522-49-4
  • Molecular Weight : 242.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011785&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011785.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart