AC5372910729 Accela ChemBio Inc.

Samarium(III) Acetate Hydrate

  • MDL number MFCD00150123
  • Cas Number : 100587-91-5
  • Molecular Weight : 327.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99.9
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011809&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011809.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart