AC5372910733 Accela ChemBio Inc.

3-Amino-5-methylpyridazine

  • MDL number MFCD10698051
  • Cas Number : 144294-43-9
  • Molecular Weight : 109.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011821&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011821.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart