AC5372910744 Accela ChemBio Inc.

Methyl 4-[N-Benzyl-N-(2-ethoxy-2-oxoethyl)amino]butanoate

  • MDL number MFCD18071032
  • Cas Number : 1256633-23-4
  • Molecular Weight : 293.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011930&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011930.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart