AC5372910745 Accela ChemBio Inc.

Methyl 4-[N-Boc-N-(2-ethoxy-2-oxoethyl)amino]butanoate

  • MDL number MFCD18071033
  • Cas Number : 1256633-34-7
  • Molecular Weight : 303.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011932&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011932.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart