AC5372910748 Accela ChemBio Inc.

Methyl 2-Amino-5-(methylsulfonyl)benzoate

  • MDL number MFCD12159812
  • Cas Number : 90610-65-4
  • Molecular Weight : 229.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011946&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY011946.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart