AC5372910758 Accela ChemBio Inc.

Ethyl 4-Fluoro-2-nitro-5-(1-pyrazolyl)benzoate

  • MDL number MFCD18071046
  • Cas Number : 1256633-41-6
  • Molecular Weight : 279.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY011999&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY011999.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart