AC5372910761 Accela ChemBio Inc.

N-Cbz-3-bromo-4-fluoroaniline

  • MDL number MFCD18071052
  • Cas Number : 1256633-39-2
  • Molecular Weight : 324.1450432

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012035&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012035.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart