AC5372910762 Accela ChemBio Inc.

N-(3-Bromo-4-fluorophenyl)methanesulfonamide

  • MDL number MFCD18071053
  • Cas Number : 1256633-26-7
  • Molecular Weight : 268.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012037&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012037.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart