AC5372910765 Accela ChemBio Inc.

5-Cyanopyridine-3-boronic Acid

  • MDL number MFCD07374882
  • Cas Number : 497147-93-0
  • Molecular Weight : 147.9271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012045&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P280-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012045.pdf
hstatements H302-H319-H335-H315
Availability details for
Quantity
  • Availability
  • Add to Cart