AC5372910775 Accela ChemBio Inc.

2-Chloro-6-methylbenzoyl Chloride

  • MDL number MFCD12025218
  • Cas Number : 89894-44-0
  • Molecular Weight : 189.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012203&attname=GoodsCode

Safety Information

signalword Danger
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012203.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart