AC5372910776 Accela ChemBio Inc.

3-Amino-5-(2-chlorophenyl)pyrazole

  • MDL number MFCD07788332
  • Cas Number : 126520-01-2
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012204&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012204.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart