AC5372910780 Accela ChemBio Inc.

N-Boc-L-pyroglutamic Acid Ethyl Ester

  • MDL number MFCD07778090
  • Cas Number : 144978-12-1
  • Molecular Weight : 257.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012242&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY012242.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart