AC5372910789 Accela ChemBio Inc.

Ethyl 2-Methyl-6-(trifluoromethyl)nicotinate

  • MDL number MFCD10688283
  • Cas Number : 380355-65-7
  • Molecular Weight : 233.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012285&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012285.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart