AC5372910793 Accela ChemBio Inc.

(2-Hydroxyphenyl)acetonitrile

  • MDL number MFCD00128130
  • Cas Number : 14714-50-2
  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012295&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012295.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart