AC5372910799 Accela ChemBio Inc.

3-(1H-Tetrazol-5-yl)benzylamine

  • MDL number MFCD09971259
  • Cas Number : 765877-97-2
  • Molecular Weight : 175.19056

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012310&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY012310.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart