AC5372910802 Accela ChemBio Inc.

4-(1H-Tetrazol-5-yl)phenol

  • MDL number MFCD05662709
  • Cas Number : 51517-88-5
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012317&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012317.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart