AC5372910804 Accela ChemBio Inc.

4-Benzyl-2-(chloromethyl)morpholine

  • MDL number MFCD02681886
  • Cas Number : 40987-25-5
  • Molecular Weight : 225.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012327&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P330+P331-P280-P305+P351+P338-P310
hcodes GHS05
MSDS http://pdf.accelachem.com/cn/msds/SY012327.pdf
hstatements H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart