AC5372910809 Accela ChemBio Inc.

4-(4-Bromo-2-chlorophenyl)morpholine

  • MDL number MFCD18651678
  • Cas Number : 1272756-07-6
  • Molecular Weight : 276.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012346&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012346.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart