AC5372910813 Accela ChemBio Inc.

3-Aminopyridazine-4-carbonitrile

  • MDL number MFCD18324663
  • Cas Number : 119581-52-1
  • Molecular Weight : 120.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012366&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012366.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart