AC5372910819 Accela ChemBio Inc.

1-Isopropyl-2-oxopyrrolidine-4-carboxylic Acid

  • MDL number MFCD02110460
  • Cas Number : 299920-47-1
  • Molecular Weight : 171.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012397&attname=GoodsCode

Safety Information

signalword Warning
pcodes P305 + P351 + P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY012397.pdf
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart