AC5372910821 Accela ChemBio Inc.

3-(1-Methyl-5-imidazolyl)benzoic Acid

  • MDL number MFCD18375276
  • Cas Number : 1261269-06-0
  • Molecular Weight : 202.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012407&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012407.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart