AC5372910823 Accela ChemBio Inc.

2-(5-Oxazolyl)benzonitrile

  • MDL number MFCD18375279
  • Cas Number : 1186127-13-8
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012410&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012410.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart