AC5372910827 Accela ChemBio Inc.

3-(2H-Tetrazol-5-yl)benzaldehyde

  • MDL number MFCD18064602
  • Cas Number : 136689-94-6
  • Molecular Weight : 174.15944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012441&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012441.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart