AC5372910835 Accela ChemBio Inc.

5-Amino-3-(2-bromophenyl)isoxazole

  • MDL number MFCD07383024
  • Cas Number : 119162-51-5
  • Molecular Weight : 239.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012490&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012490.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart