AC5372910839 Accela ChemBio Inc.

2-(2,6-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethylamine

  • MDL number MFCD18911729
  • Cas Number : 1305320-62-0
  • Molecular Weight : 365.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012524&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012524.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart