AC5372910843 Accela ChemBio Inc.

4-(4-Piperidyl)-1-butanol Hydrochloride

  • MDL number MFCD03265706
  • Cas Number : 199475-41-7
  • Molecular Weight : 193.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012562&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012562.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart