AC5372910844 Accela ChemBio Inc.

3-(Benzylamino)-3-methylbutanoic Acid

  • MDL number MFCD18375304
  • Cas Number : 113479-11-1
  • Molecular Weight : 207.26888

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012563&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012563.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart