AC5372910849 Accela ChemBio Inc.

3-(2-Amino-4-thiazolyl)benzoic Acid

  • MDL number MFCD12196529
  • Cas Number : 862254-43-1
  • Molecular Weight : 220.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012614&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY012614.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart